Learn More
Advanced Biomatrix PhotoCol Methacrylated Type I Collagen (Collagen Only)
Supplier: Advanced Biomatrix 5198100MG
Advanced BioMatrix offers PhotoCol, a purified methacrylated Type I bovine collagen kit. PhotoCol provides native-like 3D collagen gels with the unique attributes to be prepared at various concentrations and crosslinked to provide various gel stiffness. The PhotoCol kit consists of purified methacrylated Type I bovine collagen as the core component with other support reagents in the kit. Approximately 40% of the total lysine residues of the collagen molecule have been methacrylated. The collagen is extracted from bovine hide and contains a high monomer content. A bottle of 20 mM acetic acid solution is provided to solubilize the lyophilized methacrylated collagen at concentrations ranging from 3 to 8 mg/ml. The neutralization solution consists of an alkaline 10X phosphate buffered saline (PBS) solution which provides physiological salts and pH in the final mixture.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.