Learn More
Advanced Biomatrix PhotoCol-LAP Methacrylated Collagen + LAP Kit
Supplier: Advanced Biomatrix 52701KIT
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Advanced BioMatrix offers PhotoCol, a purified methacrylated Type I bovine collagen kit. PhotoColprovides native-like 3D collagen gels with the attributes to be prepared at various concentrations and crosslinked to provide various gel stiffness. The methacrylated Type I collagen is produced from telo-peptide intact bovine collagen where the collagen has been modified by reacting the free amines, primarily the -amines groups of the lysine residues as well as the a-amines groups on the N-termini. > 20% of the total lysine residues of the collagen molecule have been methacrylated. A bottle of 20 mM acetic acid solution is provided to solubilize the lyophilized methacrylated collagen at concentrations ranging from 3 to 8 mg/ml. The neutralization solution consists of an alkaline 10X phosphate buffered saline (PBS) solution. The photoinitiator consists of LAP to be formulated in 1X cell culture media or PBS which allows blue light photocrosslinking of the collagen at 405 nm.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.