Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phosphotungstic Acid, Crystal, Spectrum™ Chemical
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
Packaging:
Amber Glass Bottle
Specifications
CAS Max % | 1 |
Heavy Metals | 5e-05 |
Packaging | Amber Glass Bottle |
Quantity | 25 g |
Grade | Reagent |
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.
missing translation for 'provideContentCorrection'
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
missing translation for 'productTitle'
Spot an opportunity for improvement?
missing translation for 'provideContentCorrection'