Learn More
Getinge USA Lanceracid Rinse, Phosphoric Acid Based Solution, 2x2.5Gal
Supplier: Getinge USA LCA2.5P
An exceptional rinsing agent; LCA-P is used in the rinse cycle subsequently to our LANCERCLEAN Detergents. This phosphoric acid based solution works excellently for dealing with petroleum and other difficult soils. LCA-P is recommended for use in all glassware washers.