Learn More
Perkin Elmer US LLC Phosphorescence Sample Tube - Qty. 5
Supplier: Perkin Elmer US LLC 52126027
The Phosphorescence Sample Tube is made of synthetic fused silica, has an inner diameter of 2 mm, an outer diameter of 4 mm and is 75 mm long. This tube ships with the Low Temperature Phosphorescence Accessory (L2250136). With this part number, this tube can be ordered separately to replenish lab supplies. Qty. 5