Learn More
Electron Microscopy Sciences Phosphomolybdic Acid Orange G Solution 250 ML
Supplier: Electron Microscopy Sciences 2638606
Used in Goldner's Trichrome Method for connective tissue stains – Elastic tissue stain. Stain with a poly-acid step. See Luna, Lee G. Histopathologic Methods and Color Atlas of Special Stains and Tissue Artifacts. Johnson Printers, Downerts Grove, IL p151-152, c 1992.