Learn More
Phospho-CD247 (CD3 zeta) (Tyr142) Monoclonal Antibody (3ZBR4S), PE, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
$145.65 - $394.00
Specifications
Antigen | Phospho-CD247 (CD3 zeta) (Tyr142) |
---|---|
Clone | 3ZBR4S |
Concentration | 5 μL/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
12-247-841
|
eBioscience
12247841 |
25 Tests |
Each
for $145.65
Save 7% Reg : |
|
|||||
12-247-842
|
eBioscience
12247842 |
100 Tests |
Each for $394.00
|
|
|||||
Description
Description: This 3ZBR4S monoclonal antibody recognizes human and mouse CD247 (CD3z) when phosphorylated on Tyrosine 142. CD3 is a multi-protein complex expressed on the surface of thymocytes and T cells, a marker of the T cell lineage, and a critical component of T cell receptor (TCR) signaling. The complex contains a gamma chain, a delta chain, and two epsilon chains, which associate with the alpha/beta or gamma/delta chains of the TCR, and the zeta chain homodimer. The CD3 zeta chain is responsible for initiating the signaling cascade following engagement of the TCR. Each CD3 zeta chain contains three immunoreceptor tyrosine-based activation motifs (ITAM) that are phosphorylated by Src kinases such as Lck. Once phosphorylated, these ITAMs form docking sites for SH2 domain-containing proteins and function to recruit ZAP-70 to the TCR where it can further amplify and propagate signaling. Loss or mutation of CD3 zeta or the ITAM motifs results in impaired T cell development, signaling, and function. CD3 zeta phosphorylation occurs early in the TCR signaling cascade. It occurs immediately downstream of Lck activation, and upstream of ZAP-70, LAT, and SLP-76. Applications Reported: This 3ZBR4S antibody has been reported for use in flow cytometric analysis. Applications Tested: This 3ZBR4S antibody has been pre-diluted and tested by flow cytometric analysis of stimulated normal human peripheral blood cells. This may be used at 5 μL (0.25 μg) per test.
CD3z complex is crucial in transducing antigen-recognition signals into the cytoplasm of T cells and in regulating the cell surface expression of the TCR complex. T cell activation through the antigen receptor (TCR) involves the cytoplasmic tails of the CD3 subunits CD3 gamma, CD3 delta, CD3 epsilon and CD3 zeta. These CD3 subunits are structurally related members of the immunoglobulins super family encoded by closely linked genes on human chromosome 11. The CD3 components have long cytoplasmic tails that associate with cytoplasmic signal transduction molecules, and this association is mediated at least in part by a double tyrosine-based motif present in a single copy in the CD3 subunits. CD3 may play a role in TCR-induced growth arrest, cell survival and proliferation. The CD3 antigen is present on 68-82% of normal peripheral blood lymphocytes, 65-85% of thymocytes and Purkinje cells in the cerebellum. It is never expressed on B or NK cells. Decreased percentages of T lymphocytes may be observed in some autoimmune diseases. Diseases associated with CD3z include Immunodeficiency 25 and T-B+ Severe Combined Immunodeficiency Due To Cd3delta/Cd3epsilon/Cd3zeta.Specifications
Phospho-CD247 (CD3 zeta) (Tyr142) | |
5 μL/Test | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.2 | |
4930549J05Rik; A430104F18Rik; antigen CD3Z, zeta polypeptide; AW552088; Cd247; CD247 antigen; CD247 antigen, zeta subunit; Cd247 molecule; Cd3; CD3 antigen, zeta polypeptide; CD3 zeta; CD3 zeta chain; CD3e antigen; CD3-epsilon; Cd3-eta; CD3H; CD3Q; Cd3z; CD3Z antigen, zeta polypeptide (TiT3 complex); Cd3zeta; Cd3-zeta; CD3zeta chain; CD3-zeta/eta; FLJ18683; IMD25; T3E; T3z; T-cell antigen receptor complex, zeta subunit of CD3; T-cell receptor CD3 subunit zeta; T-cell receptor CD3, subunit zeta; T-cell receptor T3 eta chain; T-cell receptor T3 zeta chain; T-cell receptor zeta chain; T-cell surface glycoprotein CD3 zeta chain; TCR zeta chain; TCRE; Tcrk; TCRZ; TCRzeta | |
CD247 | |
IgG2b κ | |
Affinity chromatography | |
Antibody |
3ZBR4S | |
Flow Cytometry | |
PE | |
Mouse | |
Human, Mouse | |
P20963, P24161 | |
12503, 919 | |
Human CD247 peptide phosphorylated on Tyr142 | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
CD247 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.