Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phleomycin, Fisher BioReagents

Catalog Number BP2962100
Click to view available options
Quantity:
100 mg
Packaging:
Plastic Bottle
11006-33-0
C51H75N17O21S2
1326.379
QRBLKGHRWFGINE-UGWAGOLRSA-N
72511
2-[2-[2-[[2-[[4-[[2-[[6-amino-2-[3-amino-1-[(2,3-diamino-3-oxopropyl)amino]-3-oxopropyl]-5-methylpyrimidine-4-carbonyl]amino]-3-[(2R,3S,4S,5S,6S)-3-[(2S,3R,4R,5S)-4-carbamoyl-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-4,5-dihydroxy-6-(hydroxymethyl)-
CC1=C(N=C(N=C1N)C(CC(=O)N)NCC(C(=O)N)N)C(=O)NC(C(C2=CN=CN2)OC3C(C(C(C(O3)CO)O)O)(C)OC4C(C(C(C(O4)CO)O)(C(=O)N)O)O)C(=O)NC(C)C(C(C)C(=O)NC(C(=O)C)C(=O)NCCC5=NC(CS5)C6=NC(=CS6)C(=O)N)O
11006-33-0
C51H75N17O21S2
1326.379
QRBLKGHRWFGINE-UGWAGOLRSA-N
72511
2-[2-[2-[[2-[[4-[[2-[[6-amino-2-[3-amino-1-[(2,3-diamino-3-oxopropyl)amino]-3-oxopropyl]-5-methylpyrimidine-4-carbonyl]amino]-3-[(2R,3S,4S,5S,6S)-3-[(2S,3R,4R,5S)-4-carbamoyl-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-4,5-dihydroxy-6-(hydroxymethyl)-
CC1=C(N=C(N=C1N)C(CC(=O)N)NCC(C(=O)N)N)C(=O)NC(C(C2=CN=CN2)OC3C(C(C(C(O3)CO)O)O)(C)OC4C(C(C(C(O4)CO)O)(C(=O)N)O)O)C(=O)NC(C)C(C(C)C(=O)NC(C(=O)C)C(=O)NCCC5=NC(CS5)C6=NC(=CS6)C(=O)N)O

Disrupts the integrity of DNA, and blocks S-phase entry in the cell cycle

  • An antibiotic often used as selection agent to determine transformation efficiency of algae, fungi, and mammalian cells
  • Serves as a selection agent for transformed cells carrying the antibiotic-resistance gene Sh ble
  • Causes DNA fragmentation by introducting double-strand breaks in DNA and prevents cells from entering S-phase during the cell cycle
TRUSTED_SUSTAINABILITY
Type Phleomycin
Physical Form Liquid
Absorbance 0.75 to 1.05 AU at 244nm
Source Streptomyces verticillus
Odor None
Packaging Plastic Bottle
Formula Weight 1526.5
Sterile Filtered Pass Test
Quantity 100 mg
Chemical Name or Material Phleomycin
Hazard Statement WARNING!
Precautionary Statement Emergency Overview
May cause an allergic skin reaction. May cause skin, eye, and respiratory tract irritation. Limited evidence of a carcinogenic effect. Use personal protective equipment. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Get medical attention immediately if symptoms occur. Do not induce vomiting.

NFPA

Health:2
Flammability:0
Instability:0
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit