Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenytoin, USP, 98-102%, Spectrum™ Chemical
SDP

Catalog Number 18605158
Click to view available options
Quantity:
1 kg
Packaging:
Poly Pail
57-41-0
C15H12N2O2
252.27
CXOFVDLJLONNDW-UHFFFAOYSA-N
5,5-diphenylimidazolidine-2,4-dione
O=C1NC(=O)C(N1)(C1=CC=CC=C1)C1=CC=CC=C1
57-41-0
C15H12N2O2
252.27
CXOFVDLJLONNDW-UHFFFAOYSA-N
5,5-diphenylimidazolidine-2,4-dione
O=C1NC(=O)C(N1)(C1=CC=CC=C1)C1=CC=CC=C1

PH161, 57-41-0, C15H12N2O2

Spectrum™ Chemical Phenytoin, USP is an anticonvulsant or antiepileptic drug used to prevent and control seizures. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

CAS Max % 1
Loss on Drying 0.01
Packaging Poly Pail
Quantity 1 kg
Percent Purity 98 to 102%
Grade USP
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit