Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phenylthiocarbamide, Technical, 82%, Spectrum™ Chemical
Click to view available options
Quantity:
5 g
Packaging:
Amber Glass Bottle
Description
Spectrum™ Chemical Phenylthiocarbamide, Technical , also known as phenylthiourea, is used to grow transparent fish. Technical grade products supplied by Spectrum are indicative of a grade suitable for general industrial use. These materials may or may not have a Certificate of Analysis available.
Specifications
CAS Max % | 1 |
Packaging | Amber Glass Bottle |
Quantity | 5 g |
Percent Purity | 82% |
Grade | Technical |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction