Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phenol (Crystals), Fisher Chemical™
Click to view available options
Quantity:
100 g
500 g
1.5 kg
Packaging:
Amber Glass/Poison Pack
Specifications
Melting Point | 39°C |
Color | Colorless |
Density | 1.070g/cm³ |
pH | 4 to 5 |
Boiling Point | 182°C |
Assay Percent Range | ≥99.0 % |
Packaging | Amber Glass/Poison Pack |
Linear Formula | C6H5OH |
Quantity | 100 g |
Residue after Evaporation | 0.05% max. |
Show More |
Safety and Handling
Hazard Statement | DANGER! |
Precautionary Statement | Emergency Overview Combustible material. Causes burns by all exposure routes. Danger of serious damage to health by prolonged exposure. Possible risks of irreversible effects. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. Move to fresh air. If breathing is difficult, give oxygen. NFPA Health:3 Flammability:2 Instability:1 |
DOTInformation | DOT Class 6.1, : Poison |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction