Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phenol/Chloroform/Isoamyl Alcohol (125:24:1 Mixture, pH 4.3, Liq.), Fisher BioReagents™
Click to view available options
Quantity:
100 mL
Packaging:
Coated Amber Glass/Poison Pack
Description
Phenol/Chloroform/Isoamyl Alcohol (125:24:1 Mixture, pH 4.3, Liq.)
This mixture is used to purify total RNA from mixtures containing RNA, DNA, and protein.
Specifications
Specifications
Merck Index | 15, 7354 |
Percent Purity | ≥99% |
Identification | Pass Test |
Packaging | Coated Amber Glass/Poison Pack |
pH | 4.3 |
Quantity | 100 mL |
Physical Form | Liquid |
Chemical Name or Material | Phenol/Chloroform/Isoamyl Alcohol, 125:24:1 Mixture |
Safety and Handling
DOTInformation | DOT Class 6.1, : Poison |
Recommended Storage | 4°C |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction