Learn More
DSI LLC PHASEPREP™ II
Supplier: DSI LLC PHASEPREP II
PhasePrep™ II is an instrument designed to document and prepare solutions and mix solvents for HPLC mobile phases, titrations, extractions etc. PhasePrep™ prepares samples gravimetrically, rather than volumetrically, improving accuracy, increaseing your safety, and reducing waste. PhasePrep™ is GMP Compliant