Learn More
Felix Storch Inc ACCUCOLD ARG6PV Performance Pharmacy-Vaccine Refrigerator 6 Cu. Ft. with Glass Door
Supplier: Felix Storch Inc ARG6PV
ACCUCOLD ARG6PV Performance Pharmacy-Vaccine Refrigerator 6 Cu. Ft. with Glass Door with Advanced Temperature Control, cycle defrost and forced air cooling. Adjustable operating control range of 2°C to 8°C with Intelligent Microprocessor Digital Controller. Multiple Audio and Visual Alarms for Safer Temperature Monitoring. Digital display of the min/max temperature (updated in 15 minute increments). Remote alarm contacts. LED lighting, adjustable shelves, 3/8" probe hole, reversible and self closing door. Meets EPA and SNAP requirements. Eco-Friendly using hydrocarbons for a greener global footprint. Compatible with Omnicell, AcuDose, and Pyxis (not included), ADA compliant height. 2 Year Warranty Parts and Labor, 5-year compressor.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.