Learn More
Felix Storch Inc ACCUCOLD ARG15PV Performance Pharmacy-Vaccine Refrigerator 15 Cu. Ft. with Glass Door
SDP
Supplier:  Felix Storch Inc ARG15PV
Encompass_Preferred
ACCUCOLD ARG15PV Performance Pharmacy-Vaccine Refrigerator 15 Cu. Ft. with Glass Door with Advanced Temperature Control, cycle defrost and forced air cooling. Adjustable operating control range of 2°C to 8°C with Intelligent Microprocessor Digital Controller designed to provide +/- 1°C variation. Multiple Audio and Visual Alarms for Safer Temperature Monitoring. Digital display of the min/max temperature (updated in 15 minute increments). Remote alarm contacts. LED lighting, adjustable shelves, 3/8" probe hole, reversible door. Meets EPA and SNAP requirements. Eco-Friendly using hydrocarbons for a greener global footprint. Compatible with Omnicell, AcuDose, and Pyxis (not included),ADA Option available. 2 Year Warranty Parts and Labor, 5-year compressor.
Catalog No. 50-195-9415
Explore available promotions