Learn More
Aat Bioquest Phalloidin lysine (1 mg)
SDP
Supplier:  Aat Bioquest 5305
Encompass_Preferred
Phalloidin lysine is a convenient building block that can be used for developing a variety of phalloidin derivatives that may be for studying cellular structures and other biological applications. This catalog is for 1 mg.
Catalog No. 50-224-9400