Learn More
Aat Bioquest Phalloidin lysine (100 ug)
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Aat Bioquest 5304
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Phalloidin lysine is a convenient building block that can be used for developing a variety of phalloidin derivatives that may be for studying cellular structures and other biological applications. This catalog is for 100 ug.
This item is not returnable.
View return policy