Learn More
Aat Bioquest Phalloidin-California Red Conjugate (300 Tests)
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Aat Bioquest 23103
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
This red fluorescent phalloidin conjugate equivalent to Texas Red-labeled phalloindin selectively binds to F-actins with much higher photostability than the fluorescein-phalloidin conjugates. This catalog is for 300 Tests.
This item is not returnable.
View return policy