Learn More
Thermo Scientific™ Phadia™ β 2-Glycoprotein I Antibodies for Antiphospholipid Syndrome
Intended for the in vitro quantitative measurement of antibodies directed to β2-Glycoprotein I in human serum and plasma to aid in the diagnosis of antiphospholipid syndrome (APS) and to evaluate the thrombotic risk in patients with systemic lupus erythematosus (SLE).
$240.00 - $480.00
Specifications
For Use With (Application) | In-vitro quantitative measurement of Antibodies directed to beta2-Glycoprotein I in Human Serum and Plasma to aid in the diagnosis of antiphospholipid syndrome |
---|
Catalog Number | Mfr. No. | Quantity | Type | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Type | Price | Quantity & Availability | ||||
14-553-301P
|
Thermo Scientific™
14553301 |
4 x 12 Tests | EliA™ Beta2-Glycoprotein I IgM |
Pack of 48 for $480.00
|
|
||||
14-553-201P
|
Thermo Scientific™
14553201 |
4 x 12 Tests | EliA™ Beta2-Glycoprotein I IgG |
Pack of 48 for $480.00
|
|
||||
14-553-101P
|
Thermo Scientific™
14553101 |
2 x 12 Tests | EliA™ Beta2-Glycoprotein I IgA |
Pack of 24 for $240.00
|
|
||||
Description
If present in the patient's specimen, antibodies to β2-Glycoprotein I bind to their specific antigen. After washing away non-bound antibodies, enzyme-labeled antibodies against human IgG (IgM, IgA) antibodies (EliA IgG, IgM, IgA Conjugate™) are added to form an antibody-conjugate complex. After incubation, non-bound conjugate is washed away and the bound complex is incubated with a Development Solution. After stopping the reaction, the fluorescence in the reaction mixture is measured. To evaluate test results, the response for patient samples is compared directly to the response for calibrators.
- The EliA 2-Glycoprotein I IgG (IgM, IgA) Wells are coated with human β2-Glycoprotein I antigen
- The higher the responseβ value, the more specific IgG (IgM, IgA) is present in the specimen
- All conjugates are isotype-specific -- mixed conjugate is not available
Specifications
In-vitro quantitative measurement of Antibodies directed to beta2-Glycoprotein I in Human Serum and Plasma to aid in the diagnosis of antiphospholipid syndrome |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.