Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ PH111 pH/mV Bench Meter Difficult Sample Kit

Orion Lab Star PH111 pH bench meters measure pH or ORP with mV and temperature, feature a large backlit display for easy viewability, and offer small footprint to save bench space. Ideal for simplified, budget-friendly lab testing and data recording.
Supplier: Thermo Scientific™ LSTAR1118/EMD
Description
Orion Lab Star PH111 pH Meter Features
- Budget-friendly pH meter for simplified pH and ORP testing
- Display shows pH, mV and temperature simultaneously for accurate data recording
- Measure -2 to 18 pH, -2000 to +2000 mV and -5 to 105 °C or 23 to 221 °F to ensure comprehensive measurement capabilities
- Perform one to five point pH calibrations, one point ORP calibrations and one point temperature adjustment calibrations for proficient SOP compliance
- Large 5 inch backlit display makes viewing and recording measurements easy
- Small meter footprint saves valuable bench space in your lab
- Onscreen stabilizing and ready indicators help identify when readings are complete
- pH electrode condition icon helps ensure reliable electrode performance
- Date and time data stamped data and calibration logs support GLP compliance
- View active pH, ORP and temperature calibration logs with date and time stamp to ensure accurate, reproducible measurements
- Use 500-point data log with date and time stamp to record key measurements
- Output data and calibration logs to a printer or computer for long-term data storage
- Interchangeable wall plugs for US/Japan, Euro, UK/Singapore, Australia/New Zealand and China outlets
Meter and Meter Kits
- LSTAR1110 is the pH meter and stand option, which allows you to purchase your pH electrode separately
- LSTAR1115 is the standard pH meter kit option, for common pH samples such as drinking water, tap water, buffers (non-TRIS) and general purpose student use
- LSTAR116 is the easy-to-clean bio pH meter kit option, for pH samples containing TRIS buffers, proteins, sulfides, halides or other components that can interfere with silver-chloride; included refillable pH electrode is easy-to-clean and restore
- LSTAR117 is the low-maintenance bio pH meter kit option, for pH samples containing TRIS buffers, proteins, sulfides, halides or other components that can interfere with silver-chloride; included gel-filled pH electrode is low-maintenance and easy to operate
- LSTAR1118 is the difficult sample pH meter kit option, for pH samples that are thick, viscous or semi-solid including foods, beverages, wastewaters, emulsions, soils, slurries, suspended solids or sludges
Specifications
pH/mV Bench Meter Difficult Sample Kit | |
Thick, Viscous or Semi-solid Foods, Beverages, Wastewaters, Emulsions, Soils, Slurries, Suspended Solids or Sludges | |
pH, ORP, Temperature | |
0.001, 0.01, 0.1 | |
1 to 5 Points | |
0.1 mV | |
1 (ORP) | |
23°F to 221°F | |
±0.3°C (±0.5°F) | |
1 Point Offset Calibration | |
Yes | |
500 Points with Date and Time Stamp | |
Transfer Single or All Data Points to Printer or Computer | |
BNC for pH or ORP, 8-pin MiniDIN for ATC | |
CE, TUV 3-1, FCC Class A | |
198.3 x 155.2 x 61.7 mm (7.81 x 6.11 x 2.43 in.) | |
1.54 lb. | |
Universal | |
50/60 Hz |
Sure-Flow Refillable Epoxy Body pH Electrode, ATC Temperature Probe, pH 4/7/10 Buffer, Storage and Cleaning Solution, 60 mL Kit Meter with Electrode Stand, PC Cable, Power Adapter | |
pH/mV and Temperature, ORP/mV and Temperature | |
-2 to 18 | |
±0.005, ±1 LSD | |
±2000 mV with ±250 mV Offset | |
±0.2% or ±0.05% (Whichever is Greater) | |
-5°C to +105°C | |
0.1°C (0.1°F) | |
Manual, Automatic | |
LCD, Backlit | |
Non-volatile | |
Automatic with Auto-read and Timed Read Modes, Manual with Continuous Read Mode | |
Calibration Reminder Alarm | |
Computer or Printer (STARA-106) | |
IP54 | |
700 g | |
3 Years (Meter) | |
100/240 V |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title