Learn More
Cole-Parmer Oakton Waterproof pH Electrode with ATC, Single-Junction, Epoxy-Body, Gel-Filled
Supplier: Cole-Parmer 3580871
Use with the following Oakton meters: pH 300/310, pH/CON 300, and pH/CON 510. Electrode Type: Single junction, Electrode Body Material: Epoxy, Reference Junction: Gel-filled, Temperature Compensation: Yes, ATC Connector: 6-pin waterproof, Connector: BNC.