Learn More
Cole-Parmer Oakton All-in-One pH/ATC Electrode, Epoxy, Sealed, Single-Junction, 12 mm OD; 3 ft Sub-Mini/BNC
Supplier: Cole-Parmer 3581171
All-in-one combination, pH/Temp electrode, 3-ft cable, epoxy body, single-junction, gel filled, sub-miniature audio ATC, BNC, 12 mm dia.