Learn More
Bellco Glass, Inc. Mettler Toledo ph probe ¿ inpro seriesLow-maintenance ph-electrodesWith vario pin connectionMounting diameter 12mmInsertion length 425mmPh range 012Temperature range 140cPressure resistance bar up to 4 bar over pressurePressure resistance psi up t
SDP
Supplier:  Bellco Glass, Inc. 780322425
Encompass_Preferred
Mettler Toledo ph probe ¿ inpro seriesLow-maintenance ph-electrodesWith vario pin connectionMounting diameter 12mmInsertion length 425mmPh range 012Temperature range 140cPressure resistance bar up to 4 bar over pressurePressure resistance psi up t
Catalog No. 50-194-6297