Learn More
Bellco Glass, Inc. Mettler Toledo ph probe ¿ inpro seriesLow-maintenance ph-electrodesWith vario pin connectionMounting diameter 12mmInsertion length 225mmPh range 012Temperature range 140cPressure resistance bar up to 4 bar over pressurePressure resistance psi up t
Supplier: Bellco Glass, Inc. 780322225
Mettler Toledo ph probe ¿ inpro seriesLow-maintenance ph-electrodesWith vario pin connectionMounting diameter 12mmInsertion length 225mmPh range 012Temperature range 140cPressure resistance bar up to 4 bar over pressurePressure resistance psi up t