Learn More
Grainger pH Stick 25 PK Range: 0.0 to 13.9 pH
Supplier: Grainger 8ZDP7
This LAB SAFETY SUPPLY pH stick gives a quick 1-dip single-color match for each pH value. The extra long and wide strip eases use when wearing gloves and helps keep you away from potentially hazardous spills.