Learn More
Cole-Parmer Oakton pHTestr 30+ Waterproof Pocket Tester
Supplier: Cole-Parmer 3563430
Waterproof pHTestr 30 is great for outdoor applications such as taking pH measurements in lakes and streams. Designed to float in water, you can even attach a line to the housing for easy retrieval. Valox sensor housing resists most chemicals. The pHTestr 30 has a replaceable sensor for easy maintenance. pHTestr 30 purchased before Oct. 2019 use replacement sensor 35624-38. pHTestr 30 purchased after Oct. 2019 use replacemenr sensor 35634-03. Double-junction electrode sensor handles wastewater and solutions containing sulfides, heavy metals, and TRIS. Electrode sensor offers a clog-resistant Kynar junction and a recessed, glass pH bulb to protect against breakage.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.