Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ pH Paper Rolls
SCI_ED

General purpose pH papers in efficient 15' rolls

 View more versions of this product

Catalog No. S22503


Add to Cart

Description

Description

  • Convenient dispenser allows you to select the right amount of pH paper for each application
  • Easy-to-usedash (em)Dip paper in solution to be tested and match to color chart immediately
  • Highly accurate and well suited for most applications
  • Each SKU is packed in box of five clear plastic dispensers
  • Cat. No. 13-640-507 is a pH paper testing set that includes shrink wrapped pack of seven dispensers (one of each SKU)
  • Store at room temperature. Keep out of direct sunlight.

Order Info

15 ft. Roll provides about 100 tests.

Specifications

Specifications

8 to 9.5
0.5
Room Temperature
Convenient dispenser allows to select the right amount of pH paper for each application, highly accurate and well suited for most applications
100 Tests
pH Test Paper
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit