Learn More
World Precision Instrument pH MicroImplant Fiber Optic Implantable Sensor
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: World Precision Instrument 502124
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
The pHOptica micro system is a single channel pH system for use with fiber optic micro sensors.