Learn More
CTL Scientific Supply Corp PH INDICATOR STRIPS 7.9-9.8
Supplier: CTL Scientific Supply Corp 92170
Non-bleeding indicator strips. Gradation of scale: 7.9 - 8.3 - 8.6 - 8.9 - 9.1 - 9.4 - 9.8 100 tests/box