Learn More
Broadley James Corporation Soaker Bottles,4-Pack of Soaker Bottles for 12mm probes, 4 Pack
Supplier: Broadley James Corporation AM109012
Soaker Bottles,4-Pack of Soaker Bottles for 12mm probes,Protection for any 12 mm sensor during transportation or storage,The o-ring fits securely against the electrode body,sealing the solution from the atmosphere to minimize evaporation