Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisher Science Education™ pH Buffer Solution
Moderate Hazard
$8.80
Specifications
CAS | 1310-73-2, 7778-77-0, 110-44-1, 7732-18-5 |
---|---|
Density | 1.000 |
Physical Form | Liquid |
Quantity | 500 mL |
Catalog Number | Quantity | Packaging | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Quantity | Packaging | Price | Quantity & Availability | |||||
S25849
|
500 mL | pH 10.0: Blue |
Each for $8.80
|
|
|||||
S25849A
|
500 mL | pH 4.0: Red |
Each for $8.80
|
|
|||||
S25849B
|
500 mL | pH 7.0: Yellow |
Each for $8.80
|
|
|||||
Specifications
1310-73-2, 7778-77-0, 110-44-1, 7732-18-5 | |
1.000 | |
Liquid | |
500 mL |
Safety and Handling
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title