Learn More
Inorganic Ventures pH 4 CALIBRATION STD 1L (Inorganic Ventures)
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Inorganic Ventures PH41L
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Inorganic Ventures' pH 4 Standard is a 1L certified reference material set in a water matrix for stability. This standard is Traceable to NIST and manufactured in accordance with our stringent Quality Assurance guidelines. One year Expiration Date.
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.