Learn More
Molecular Dimensions PGA SCREEN (PREMIXED)
SDP
Supplier:  Molecular Dimensions MD150
Encompass_Preferred
Protein Crystallography. PGA SCREEN (96 x 10mL): Poly- Gamma -glutamic acid polymer, PGA, is the first of a new breed of chemical polymers. PGAs can be used at very low concentrations and with classical precipitants, thus decreasing precipitant necessary for crystal appearance and growth.
Catalog No. NC9945797
Explore available promotions