Learn More
Electron Microscopy Sciences PetriOrienter, 6-sector pie
SDP
Supplier:  Electron Microscopy Sciences 7049807
Encompass_Preferred
The Petri Orienter creates a consistent area of reference on your petri dish and sharp characters and lines allow for fast and easy position recognition.
Divisions: 6-sector pie
Catalog No. 50-345-54