Learn More
Nelson Jameson Company E.coli/Coliform Count Plates 50/pack
Supplier:  Nelson Jameson Company 4603404 BC
Encompass
E.coli/Coliform Count Plates identify both E.coli and other coliforms. With this one easy test, you'll have confirmed results in 24 to 48 hours. Maximize productivity by increasing your laboratory efficiency with 3M™ Petrifilm™ Plates. SPREADER INCLUDED.
Catalog No. NC0940366