Learn More
Research Products International Corp PetriStickers, 70 Square Grid, 3 Inch Diameter, 36 per Package
Supplier: Research Products International Corp 24701070
PetriStickers allow for petri dishes to be labeled quickly, neatly and reliably.Made with water-resistant ink, substrate and adhesives, they will remain firmly attached at temperatures from 42? to 4?C.Simply remove the label's protective backing and apply it to the back of a petri dish.After labeling, the dish can be held in a comfortable position for streaking or colony transfer, stored in an incubator or refrigerator, and photographed.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.