Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisherbrand™ Petri Dishes with Clear Lid
Feature a new and improved design and packaging
Description
- Made from medical-grade, gamma sterilized polystyrene
- Ventilation ribs on the lid allow for air circulation
- Manufactured in a state-of-the-art cleanroom facility
- Packaging features a UPS™ shippable outer carton to prevent damage
- Product sleeve is made from a strong polymer to prevent bag holes
- Dishes packed in easy-open product sleeve with tear notch
- Product sleeves are vacuum sealed to prevent scuffing and other damage to dishes
- Complete manufacturing traceability with use of both the lot number on the case label and the lot ID number on the product sleeve
Specifications
Stackable | |
Polystyrene | |
Raised Ridge | |
15 mm |
Clear | |
Round | |
60 x 15 mm | |
60 mm |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction