Learn More
Market Lab MarketLab Media Plate Holders, 100 mm, 1 each
Supplier: Market Lab ML7009
Small footprint ideal for limited benchtop or wall space. Will not bend or crack inside incubator. Constructed of durable acrylic. For 100mm Media Plates, 4"W x 4"D x 9"H.