Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Wheaton™ PET Liquid Scintillation Vials
For optimal performance in the laboratory. Wheaton™ PET Liquid Scintillation Vials offer low permeability to solvents and minimal background counts.
Supplier: Wheaton™ 986744
Description
- With caps attached
- Available linerless or with foamed polyethylene liner
Specifications
27 mm | |
20 mL | |
Polypropylene | |
61 mm | |
Scintillation Vial |
PET | |
Linerless | |
Screw Cap | |
1000 Pack | |
22-400 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title