Learn More
Thermo Scientific Peristaltic Pump Tubing for iCAP™ Q/Qnova Series ICP-MS Systems, 12/pk
Supplier:  Thermo Scientific 1320050
Encompass
PVC peristaltic pump tubing - ID 0.508 mm - orange/yellow (3 bridges, flared end). 12 tubings.
Catalog No. NC0587270