Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex L/S Variable-Speed, Compact, Drive, 35 to 200 rpm; 115/230 VAC
Supplier: AVANTOR MASTERFLEX (VWR LLC) MFLX7720020
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
These compact drives are a perfect when space is limited, as they fit just about anywhere. The variable-speed design makes them ideal for when you need to pump fluid at a higher rate. Drives accept one Easy-Load or standard pump head. Use drives with L/S Precision tubing¿accepts sizes L/S 13, L/S 14, L/S 16, L/S 25, and L/S 17. Only silicone and C-Flex tubing formulations are recommended for use with L/S 25 and L/S 17 sizes.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.