Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Peri-xanthenoxanthene, 95%, Thermo Scientific™

Catalog Number AC454892500
Click to view available options
Quantity:
250 mg
1 g
191-28-6
C20H10O2
282.3
AMDQVKPUZIXQFC-UHFFFAOYSA-N
dinaphthylene dioxide, peri-xanthenoxanthene, dinaphthalene dioxide, xantheno 2,1,9,8-klmna xanthene
240798
C1=CC2=C3C(=C1)OC4=C5C3=C(C=C2)OC6=CC=CC(=C65)C=C4
191-28-6
C20H10O2
282.3
AMDQVKPUZIXQFC-UHFFFAOYSA-N
dinaphthylene dioxide, peri-xanthenoxanthene, dinaphthalene dioxide, xantheno 2,1,9,8-klmna xanthene
240798
C1=CC2=C3C(=C1)OC4=C5C3=C(C=C2)OC6=CC=CC(=C65)C=C4
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Quantity 250 mg
Formula Weight 282.3
Percent Purity 95%
Chemical Name or Material Peri-xanthenoxanthene
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May cause an allergic skin reaction.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing.
In case of inadequate ventilation wear respiratory protection.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation or rash occurs: Get medical advice/attenti

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit