Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Eisco™ Performance Plastic Chemical Wash Bottles with 2 Color Labels
Click to view available options
Capacity:
500 mL
1000 mL
Chemical Label:
Acetone
Distilled Water
Ethanol
Isopropanol
Methanol
Sodium Hypochlorite (Bleach)
Quantity:
1/Pk.
2/Pk.
6/Pk.
Description
- Chemical specific wash bottles with color coded caps and labels
- Performance plastic is durable yet extremely responsive
- Made of low-density polyethylene (LDPE)
- Easy to read labeling, including chemical specific information and safety warnings
- Self-venting design to eliminate leaks and dripping
- Wide mouth for easy filling
- Innovative flex ink won't fade, crack, or peel
- Sodium Hypochlorite (bleach) wash bottles are opaque (white) to protect contents from UV light rays
- Other bottles are clear/transparent
- Bottle labels comply with the Globally Harmonized System of Classification and Labeling of Chemicals (GHS)
- Bottle Labels also comply with OSHA's HazCom 2012 requirements and Hazard Communication Standard 29 CFR 1910.1200
Specifications
Closure Type | Screw Cap |
Closure Size | 5 cm |
Chemical Label | Methanol |
Product Type | Wash Bottle |
Capacity | 500 mL |
Capacity (English) | 2 c |
Color | Clear |
Material | LDPE |
No. per Pack | 1 |
Quantity | 1/Pk. |
Show More |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction