Learn More
Thermo Scientific™ Perfluoroacylimidazoles (TFAI and HFBI) Acylation Reagents
Conduct acylation of hydroxyl groups and primary and secondary amines with the Thermo Scientific™ Perfluoroacylimidazoles (TFAI and HFBI).
Supplier:  Thermo Scientific™ TS44211
 View more versions of this product
Catalog No. PI44211
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Description
Description
Conduct acylation of hydroxyl groups and primary and secondary amines with the Thermo Scientific™ Perfluoroacylimidazoles (TFAI and HFBI). Compared to acid chlorides and anhydrides, the reactions of fluorinated acylimidazoles are smooth and quantitative, and produce no acid byproducts. The principal byproduct is imidazole, which is relatively inert. The volatile and stable fluoroacyl derivatives are ideal for FID and electron-capture detection (ECD) techniques.
Applications
Used in bifunctional derivatization schemes and in exchange reactions in which TMS derivatives are converted to HFB derivatives.
Hydroxyl groups of catecholamines are derivatized with TMSI, followed by conversion of the amines to acylamides with HFBI.
Tryptamine and metabolites present in spinal fluid have been analyzed by ECD using HFBI.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
FID, ECD
-20°C
1-(2,2,3,3,4,4,4-Heptafluoro-1-oxobutyl)-1 H-imidazole
HFBI; 5 g
Combustible, Irritant
5 g
98 to 100%
Product Suggestions
Product Suggestions
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only. Not for use in diagnostic procedures.