Learn More
Electron Microscopy Sciences Perfect Handle Only
Supplier: Electron Microscopy Sciences 70616HD
Made from Nickel alloy, 3 mm in length with 1/10 mm divisions. The Scale fits the Perfect Handle (70616-HD). The Perfect Scale comes complete with one scale and a handle.