Learn More
Glas Col LLC Control
Supplier: Glas Col LLC 104A RL3312
Percentage Timerwhich pulses full line voltage to each postion according to dial setting. For 3 place RJ (jigh temp) or RX (low temp) Combo Mantle 120V, 12A