Learn More
NEOGEN Peptone Water, 500 g
SDP
Supplier:  NEOGEN 700003239
Encompass_Preferred
Peptone Water is used for the cultivation of non-fastidious microorganisms, indole testing, and as a basal medium for carbohydrate fermentation studies. Peptone Water is not intended for use in the diagnosis of disease or other conditions in humans. A general purpose growth medium that can be used as a base for carbohydrate fermentation studies. The medium has a high level of tryptone making it suitable for use in the indole test.For more information visit https://www.neogen.com/solutions/Microbiology/peptone-water
Catalog No. 50-201-5435
Explore available promotions