Learn More
Cole-Parmer Oakton Rugged pH Electrode,12x110mm, Glass Body, Double Junction, Refill
Supplier: Cole-Parmer 5551038
Cole-Parmer high-performance electrodes are available in a full range of materials, junctions, and sizes to suit your application requirements. New formulation provides additional stability and superior reproducibility over entire pH range. Fill solution purchased seperately, 05992-47 for single junction, 05992-48 for double juncion electrodes