Learn More
Electron Microscopy Sciences Pen Vac™ System
Supplier: Electron Microscopy Sciences 71914
Pen-Vac is a new improved way to handle small, flat surface objects. Beside the electronics assembly industry, jewelers, model builders etc. Pen-Vac is ideal for EM work as well. It can be used to handle grids, pick up stubs, align membranes, work with glass slides, cover slips and much more. Holds up to one minute.
Pen-Vac™ comes with a variety of straight and angle vacuum probes complete with a vacuum cap. The stainless steel needle portion of the probes are one-half inch long. Lifts up to 50 grams.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.