Learn More
3M™ PELTOR™ Push-To-Talk (PTT) Adapter, Motorola Turbo, NATO Wiring
Use 3M™ PELTOR™ Push-To-Talk (PTT) Adapters optimize the placement of the PTT button for convenient activation
Supplier: 3M™ 7010384262
Description
- For use with 3M™ PELTOR™ COMTAC™™ Protective Communication Headsets and compatible two-way radios
- Ambidextrous design allows for easy use by either the right or left hand
- Mounting clip can be rotated 360 degrees
- Design allows for either belt or shoulder mounting
- Button profile and tactile feedback suitable for gloved hands
- Optional shroud protects PTT button from accidental activation, for instance when laying in the prone position
- NEXUS™ TP-120 connection serves as a break-away connector for removal in emergency situations
- Curly cable downlead with radio specific connector. To determine the appropriate PTT adapter for your two-way radio, please visit 3M.com/PeltorAdapters
- NATO wired
Specifications
Adapter | |
For use with 3M™ PELTOR™ COMTAC™™ Protective Communication Headsets and Compatible Two-way Radios | |
FL5063-02 | |
IP65 |